15-Ketoeicosa-6E,8Z,11Z,13E-tetraenoic acid ≥95% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES
Gefahr

BMLK0150050
ENZOBMLK0150050EA 225 EUR
ENZOBMLK0150050
15-Ketoeicosa-6E,8Z,11Z,13E-tetraenoic acid ≥95% (durch HPLC)
15-Ketoeicosa-6E,8Z,11Z,13E-tetraenoic acid

Biosynthesis via 15-OH prostaglandin dehydrogenase from lung and liver microsomes and by a nonenzymatic process.

Lagertemperatur: Ultra-Tiefkühlschrank (–80°C)
CAS-Nummer: 81416-72-0
UN: 1170
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ea5566f1fb21f9d24fdc30ac2b81372cd7784b0&vid=c68e4ebf7159a6ba9e05e62351eb3ebb32086878&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥95% (HPLC)
Concentration (for 50 µg size) 50 µg/ml for 50 µg size
Concentration (for 1 mg size) 1 mg/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR