Minoxidil sulphate ≥98%
Lieferant: ENZO LIFE SCIENCES
|
Achtung
|
K⁺ channel activator.
A selective ATP-sensitive K⁺ (KATP) channel opener. It is the active metabolite of minoxidil and is a potent (IC₅₀=0.14 µM) vascular smooth muscle relaxant. Antihypertensive. Stimulates renin secretion.
Soluble in DMSO (25 mg/ml).
Formel:
C₉H₁₅N₅O₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD06248907 CAS-Nummer: 83701-22-8 |
Spezifikation Testergebnisse
Purity | ≥98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...