Valinomycin ≥97% (durch TLC), weißer Feststoff
Lieferant: ENZO LIFE SCIENCES
BMLKC1400025
ENZOBMLKC1400025EA
89.1
EUR
ENZOBMLKC1400025
Valinomycin ≥97% (durch TLC), weißer Feststoff
Valinomycin
Potassium ionophore.
- Isolated from Streptomyces fulvissimus
- White crystalline solid
Induces mitochondrial swelling and apoptosis in CHO cells and other cell lines. It antagonises endothelin-induced vasoconstriction (IC50=0,3 µM).
Soluble in 100% ethanol (50 mg/ml), acetic acid, chloroform, DMSO or ether
Bestellinformation: Shipped at ambient temperature
Formel:
C₅₄H₉₀O₁₈N₆ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00005114 CAS-Nummer: 2001-95-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...