4,4-Pentamethylene-2-pyrrolidone ≥95%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 2-Azaspiro[4.5]decan-3-one, Gabapentin-lactam, 3-Azaspiro[4.5]decan-2-one

BMLKC1600250
ENZOBMLKC1600250EA 262 EUR
ENZOBMLKC1600250
4,4-Pentamethylene-2-pyrrolidone ≥95%
4,4-Pentamethylene-2-pyrrolidone

Gabapentin-lactam but not gabapentin reduces protein aggregates and improves motor performance in a transgenic mouse model of Huntington’s disease. It opens mitochondrial ATP-dependent potassium channels.

Formel: C₉H₁₅NO
MDL: MFCD00177938
CAS-Nummer: 64744-50-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7081be438aba8ae55cf702da011f8e40dcdb1a1c&vid=37bf94fcd826795851ac7b9965d24129769df5fd&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR