Caspase assay buffer
Lieferant: ENZO LIFE SCIENCES
BMLKI1110020
ENZOBMLKI1110020EA 49.5 EUR
ENZOBMLKI1110020
Caspase assay buffer
Puffer Assay-Puffer
50 mM HEPES, pH 7,4, 100 mM NaCl, 0,1% CHAPS, 10 mM DTT, 1 mM EDTA, 10% glycerol.