Nuclear extract from HeLa cells
Lieferant: ENZO LIFE SCIENCES
BMLKI1400100
ENZOBMLKI1400100EA 165 EUR
ENZOBMLKI1400100
Nuclear extract from HeLa cells
Zellen
Prepared by high salt extraction of HeLa nuclei (human cervical cancer cell line), this extract is a rich source of HDAC activity, useful for inhibitor screening or immunoprecipitation of HDAC containing complexes. Typically, 0,5 µl or less per well is sufficient for assay with the HDAC FLUOR DE LYS® fluorometric activity assay.