Natriumdodecylsulfat (SDS)

Lieferant: ENZO LIFE SCIENCES
Gefahr

BMLKI3410012
ENZOBMLKI3410012EA 49.5 EUR
ENZOBMLKI3410012
Natriumdodecylsulfat (SDS)
Natriumdodecylsulfat (SDS)

Component of the 20S proteasome drug discovery assay.

Formel: CH₃(CH₂)₁₁OSO₃Na
Lagertemperatur: Kühlschrank
MDL: MFCD00036175
CAS-Nummer: 151-21-3
UN: 2926
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR