HDAC assay buffer II
Lieferant: ENZO LIFE SCIENCES
BMLKI4220020
ENZOBMLKI4220020EA
48.4
EUR
ENZOBMLKI4220020
HDAC assay buffer II
Puffer
Assay-Puffer
50 mM Tris/Cl, pH 8.0, 137 mM NaCl, 2.7 mM KCl, 1 mM MgCl2, 1 mg/ml BSA.