Cathepsin S Assay Buffer
Lieferant: ENZO LIFE SCIENCES
BMLKI4310020
ENZOBMLKI4310020EA
52.8
EUR
ENZOBMLKI4310020
Cathepsin S Assay Buffer
Puffer
Assay-Puffer
50mM potassium phosphate buffer, pH 6,5, 50 mM NaCl, 2 mM EDTA, 0,01% Triton X-100®, 0,5 mM DTT.