FLUOR DE LYS®-succinyl green, desuccinylase substrate 5 mM in DMSO (synthetisch) , 95% (durch HPLC)

Lieferant: Enzo Life Sciences
Achtung

BMLKI5910050
ENZOBMLKI5910050EA 252 EUR
ENZOBMLKI5910050
FLUOR DE LYS®-succinyl green, desuccinylase substrate 5 mM in DMSO (synthetisch) , 95% (durch HPLC)
FLUOR DE LYS®-Succinyl Green, Desuccinylase Substrate

The FLUOR DE LYS®-Succinyl green, desuccinylase substrate offers a highly sensitive and convenient alternative to radiolabeled, succinylated histones or peptide/HPLC methods for the assay of desuccinylases such as SIRT5.


The substrate has longer wavelength excitation and emission. This avoids interference by quenching or fluorescence from compounds absorbing and emitting in the near UV and blue range.


Can be used for the study of desuccinylase kinetics, regulation, and inhibitor sensitivity. Ideal for drug discovery and HTS applications.

Bestellen

Spezifikation Testergebnisse

Concentration 5 mM
Source Synthetic
Absorbance maximum 470 - 500 nm
Emission maximum 520 - 550 nm
Purity ≥95% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR