1-O-Hexadecyl-2-N-methylcarbamyl-sn-glycero-3-phosphocholine ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Carbamyl-PAF

BMLL1200005
ENZOBMLL1200005EA 385 EUR
ENZOBMLL1200005
1-O-Hexadecyl-2-N-methylcarbamyl-sn-glycero-3-phosphocholine ≥98% (durch TLC)
1-O-Hexadecyl-2-N-methylcarbamyl-sn-glycero-3-phosphocholine

PAF receptor activator.


Biologically potent, non-metabolisable analog of platelet-activating factor (PAF). Induces half-maximal PMNL degranulation response at 16 nM and binds stereospecifically to high and low affinity PAF receptors. Completely resists inactivation by acetylhydrolase.


Soluble in 100% ethanol.

Formel: C₂₆H₅₅N₂O₇P
Molecular Weight: 538,707 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00065891
CAS-Nummer: 91575-58-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90acfc8f4a07ce60727394a75b7fd0e26a37aecf&vid=b0f35ebf530251d070308c9567dea1e05905ff73&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR