1-O-Palmitoyl-2-O-acetyl-sn-glycero-3-phosphocholine ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLL1340001
ENZOBMLL1340001EA 64.9 EUR
ENZOBMLL1340001
1-O-Palmitoyl-2-O-acetyl-sn-glycero-3-phosphocholine ≥98% (durch TLC)
1-O-Palmitoyl-2-O-acetyl-sn-glycero-3-phosphocholine

PAF receptor ligand.


The predominant PAF species produced by stimulated endothelial cells. 1-Acyl-PAF induces IL-6 release in rat anterior pituitary cells in vitro. The compound also displays protective effects in oxidative stress.


Soluble in DMSO (25 mg/ml) or ethanol (25 mg/ml).

Formel: C₂₆H₅₂NO₈P
Molecular Weight: 537,675 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00798242
CAS-Nummer: 84062-61-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89889ef5f1bb115853b34e583c05e0564aebbc04&vid=b47a71ab698ca2d65ed1c7f197ae4c46be24cce9&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR