Leukotriene E4-20,20,20-d3 ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
Synonyme:
LTE4-d3
BMLLE5040050
ENZOBMLLE5040050EA
467
EUR
ENZOBMLLE5040050
Leukotriene E4-20,20,20-d3 ≥98% (durch HPLC)
Leukotriene E4-20,20,20-d3
Prostanoid receptor ligand.
Formel:
C₂₃H₃₄D₃NO₅S Molecular Weight: 442,6 g/mol |
UN:
1992 ADR: 3,II |
Spezifikation Testergebnisse
Source | Synthetic |
Formulation | Solution in methanol/ammonium acetate buffer, 70:30, pH 5.6 |
Concentration | 50 µg/ml |
Purity | ≥98% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...