L-NASPA ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: N-Palmitoyl-L-serine phosphoric acid

BMLLP1010001
ENZOBMLLP1010001EA 84.7 EUR
ENZOBMLLP1010001
L-NASPA ≥98% (durch TLC)
L-NASPA

L-NASPA is an antagonist of LPA-induced platelet aggregation and chloride conductance in Xenopus oocytes. In human breast carcinoma MDA-MB231 cells it is a potent LPA agonist.


Soluble in DMSO (10 mg/ml)or ethanol (10 mg/ml).

Formel: C₁₉H₃₈NO₇P·H₂O
Lagertemperatur: Ultra-Tiefkühlschrank (–80°C)
CAS-Nummer: 155915-46-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=772ba9d6a058abb44874bd967c2b7acc681648a1&vid=5efc5aed3d26231fc6cac7be280220fc012aaee7&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR