Phosphatidic acid, dioleoyl ≥98% (durch GC), weißes Pulver

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: 1,2-Dioleoyl-sn-glycero-3-phosphate, sodium salt, (Z)-(R)-3-(Phosphonooxy)propane-1,2-diyl dioleate sodium salt

BMLLP1040010
ENZOBMLLP1040010EA 165 EUR
ENZOBMLLP1040010
Phosphatidic acid, dioleoyl ≥98% (durch GC), weißes Pulver
Phosphatidic acid, dioleoyl

PKC activator


A mitogenic lipid phosphate produced via the action of phospholipase D. Mediates agonist-dependent Raf-1 translocation and activation of the MAP kinase cascade. Activates PKCζ.


Soluble in chloroform.

Formel: C₃₉H₇₃O₈P·Na
Molecular Weight: 724 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00674303
CAS-Nummer: 108392-02-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68f5ec6748eb0fac38567a400596c1c607fd3fc8&vid=4da5a649048b130f4e0345efc58bd58e49b19ce9&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR