Benzamil hydrochloride ≥98% (durch HPLC), gelber Feststoff
Lieferant: ENZO LIFE SCIENCES
|
Achtung
|
Synonyme:
N-(Benzylamidino)-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride, Benzamil HCl, Benzylamiloride hydrochloride, 3,5-Diamino-N-(N-benzylcarbamimidoyl)-6-chloropyrazine-2-carboxamide hydrochloride
BMLNA1030050
ENZOBMLNA1030050EA
271
EUR
ENZOBMLNA1030050
Benzamil hydrochloride ≥98% (durch HPLC), gelber Feststoff
Benzamil hydrochloride
Na⁺ channel blocker
Derivative of amiloride (Prod. No. BML-CA200). Potent sodium channel blocker.
Soluble in water.
Formel:
C₁₃H₁₄ClN₇O·HCl Molecular Weight: 356,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00069219 CAS-Nummer: 161804-20-2 |
Spezifikation Testergebnisse
Purity | ≥98% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...