Ketotifenfumarat ≥99%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 4-(1-Methylpiperidin-4-ylidene)-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-10(9H)-one fumarate

BMLNH1060001
ENZOBMLNH1060001EA 84.7 EUR
ENZOBMLNH1060001
Ketotifenfumarat ≥99%
Ketotifen fumarate

Histamine receptor ligand


Selective histamine H1 antagonist. Inhibits histamine release from monocytes and lymphocytes at 0.1-10 µM.

Formel: C₁₉H₁₉NOS
Molecular Weight: 425,50 g/mol
MDL: MFCD00079394
CAS-Nummer: 34580-14-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe571cc4642aff472ec3f60c9060e617788d015b&vid=77a0d0315bf1441137ff819b434fca1a4fffd5d8&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR