HOE-140 ≥95% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Icatibant, ((2S,3aS,7aS)-1-((R)-2-(((S)-2-(2-((2S,4R)-1-(D-Arginyl-L-arginyl-L-prolyl)-4-hydroxypyrrolidine-2-carboxamido)acetamido)-3-(thiophen-2-yl)propanoyl)-L-seryl)-1,2,3,4-tetrahydroisoquinoline-3-carbonyl)octahydro-1H-indole-2-carbonyl)-L-arginine

BMLNK1040001
ENZOBMLNK1040001EA 324 EUR
ENZOBMLNK1040001
HOE-140 ≥95% (durch HPLC)
HOE-140

A potent and selective bradykinin B2 antagonist.


HOE-140 is a useful tool for discrimination of the physiological roles of bradykinin B2 versus B1 receptors. Displays a protective effect in myocardial reperfusion injury.


Soluble in water.

Formel: C₅₉H₈₉N₁₉O₁₃S
Molecular Weight: 1304,5 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 130308-48-4

Bestellen

Spezifikation Testergebnisse

Purity ≥95% (HPLC)
Identity Determined by MS
Sequence D-Arg-Arg-Pro-Hyp-Gly-Thi-Ser-Tic-Oic-Arg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR