N-Desmethyl clozapine ≥99% (durch HPLC), gelber Feststoff
Lieferant: ENZO LIFE SCIENCES
|
Achtung
|
Synonyme:
Norclozapine
BMLNS1060025
ENZOBMLNS1060025EA
467
EUR
ENZOBMLNS1060025
N-Desmethyl clozapine ≥99% (durch HPLC), gelber Feststoff
N-Desmethyl clozapine
Serotonin receptor ligand.
Major metabolite of clozapine. Potent serotonin 5-HT1c receptor antagonist.
Soluble in ethanol (25 mg/ml) or DMSO (>44 mg/ml).
Formel:
C₁₇H₁₇ClN₄ Molecular Weight: 312,8 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00210189 CAS-Nummer: 6104-71-8 |
Spezifikation Testergebnisse
Purity | ≥99% (HPLC) |
Identity | Determined by ¹H-NMR |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...