9-Methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-1,2,3,9-tetrahydro-4H-carbazol-4-on ≥98% (durch NMR), weißer Feststoff
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
Synonyme:
Ondansetron
BMLNS1450050
ENZOBMLNS1450050EA
431
EUR
ENZOBMLNS1450050
9-Methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-1,2,3,9-tetrahydro-4H-carbazol-4-on ≥98% (durch NMR), weißer Feststoff
9-Methyl-3-((2-methyl-1H-imidazol-1-yl)methyl)-2,3-dihydro-1H-carbazol-4(9H)-one
Serotonin receptor ligand
A serotonin 5-HT3 receptor antagonist. Clinically effective anti-emetic especially in chemotherapy-induced nausea.
Soluble in DMSO (10 mg/ml warm).
Formel:
C₁₈H₁₉N₃O Molecular Weight: 293,36 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00833882 CAS-Nummer: 99614-02-5 |
Spezifikation Testergebnisse
Purity | ≥98% (NMR) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...