Melatonin ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
Endogenous agonist at melatonin MT1 and MT2 receptors. Displays antioxidant and antiapoptotic effects in numerous cell types as well as beneficial effects in various neurodegenerative diseases. It protects neuronal cells from amyloid beta-mediated oxidative damage and increases survival in an Alzheimer’s disease transgenic mouse model. Melatonin directly inhibits mitochondrial permeability transition pore (IC50 = 0.8M) and reduced the loss of neurons in a rat model of transient brain ischemia.
- Ultra-pure
White to off-white powder.
Soluble in dilute aqueous acid, methanol or 100% ethanol.
Formel:
C₁₃H₁₆N₂O₂ Molecular Weight: 232,28 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00005655 CAS-Nummer: 73-31-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...