[Ser25]-Protein kinase C (19-31)
Lieferant: ENZO LIFE SCIENCES
BMLP1110001
ENZOBMLP1110001EA 139 EUR
ENZOBMLP1110001
[Ser25]-Protein kinase C (19-31)
Proteine und Peptide
PKC substrate
Derived from the PKC pseudosubstrate sequence but with an Ala to Ser-25 substitution which makes it a good substrate (Km=0,2 µM) rather than an inhibitor. The resulting sequence is RFARKGSLRQKNV.