OmniMMP™ fluorogenic control
Lieferant: ENZO LIFE SCIENCES
BMLP1270001
ENZOBMLP1270001EA
92.4
EUR
ENZOBMLP1270001
OmniMMP™ fluorogenic control
Proteine und Peptide
Reference peptide for OmniMMPTM fluorogenic substrate (BML-P126) and TACE (ADAM17) fluorogenic substrate (BML-P132). Ex.: 328 nm, Em.: 393 nm.
The following Ex/Em have also been used: 320 to 340/400 to 420.