H-Arg-AMC·2HCl
Lieferant: ENZO LIFE SCIENCES
Synonyme:
L-Arginine 7-amido-4-methylcoumarin dihydrochloride
BMLP1350025
ENZOBMLP1350025EA
141
EUR
ENZOBMLP1350025
H-Arg-AMC·2HCl
H-Arg-AMC·2HCl
Substrate for cathepsin H and aminopeptidases
Fluorogenic substrate for Cathepsin H (Km=150 µM) and aminopeptidases. Ex.: 380 nm, Em.: 460 nm, although the following Ex/Em can also be used: 355,375/440,450. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144).
Formel:
C₁₆H₂₁N₅O₃·2HCl Molecular Weight: 404,3 g/mol |
MDL:
MFCD00155578 CAS-Nummer: 113712-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...