(S)-2-Amino-5-guanidino-N-(4-nitrophenyl)pentanamide dihydrochloride ≥99%

Lieferant: ENZO LIFE SCIENCES

Synonyme: L-Arginine 4-nitroanilide dihydrochloride, H-Arg-pNA·2 HCl, L-Arginine p-nitroanilide dihydrochloride, H-Arg-pNA·2HCl

BMLP1360025
ENZOBMLP1360025EA 52.8 EUR
ENZOBMLP1360025
(S)-2-Amino-5-guanidino-N-(4-nitrophenyl)pentanamide dihydrochloride ≥99%
(S)-2-Amino-5-guanidino-N-(4-nitrophenyl)pentanamide dihydrochloride

Aminopeptidase substrate


Chromogenic substrate for Cathepsin H and aminopeptidases. Release of pNA is monitored at 405-410 nm. This substrate is useful for inhibitor screening and kinetic analysis. Also available: calibration standard pNA (BML-KI145).

Formel: C₁₂H₂₀Cl₂N₆O₃
Molecular Weight: 367,231 g/mol
MDL: MFCD00058249
CAS-Nummer: 40127-11-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR