Suc-Ala-Ala-Pro-Phe-pNA
Lieferant: ENZO LIFE SCIENCES
BMLP1410025
ENZOBMLP1410025EA
90.2
EUR
ENZOBMLP1410025
Suc-Ala-Ala-Pro-Phe-pNA
Proteine und Peptide
Cathepsin substrate
Chromogenic substrate cleaved by cathepsin G, subtilisins, chymotrypsin, chymase, and cyclophilins, but not neutrophil elastase. Release of pNA is monitored at 405 to 410 nm. This substrate is useful for inhibitor screening and kinetic analysis. Also available: calibration standard pNA (BML-KI145).