Z-GPR-Amc
Lieferant: ENZO LIFE SCIENCES
BMLP1420020
ENZOBMLP1420020EA
381
EUR
ENZOBMLP1420020
Z-GPR-Amc
Proteine und Peptide
Cathepsin K substrate
Fluorogenic substrate specific for cathepsin K, among other cathepsin family members. This substrate is also cleaved by granzyme A, thrombin, tryptase, trypsin, and kallikreins. Ex.: 365 nm, Em.: 440 nm, although the following Ex/Em can also be used: 355,380/450,460. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144).