Mca-APK(Dnp)
Lieferant: ENZO LIFE SCIENCES
BMLP1630001
ENZOBMLP1630001EA 139 EUR
ENZOBMLP1630001
Mca-APK(Dnp)
Proteine und Peptide
ACE substrate
Fluorogenic substrate for angiotensin converting enzyme-2 (ACE2), a zinc-dependent carboxypeptidase with 42% amino acid sequence identity to ACE1. Unlike Mca-YVADAPK(Dnp), this substrate is not cleaved by caspases. Mca fluorescence is quenched by the Dnp group until cleavage (at Pro-Lys) separates them. Ex: 328 nm, Em: 393 nm, although the following Ex/Em have also been used: 320-340/400-420. This highly quenched (99.6%) substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic Mca control peptide (BML-P127).