Z-VVR-AMC
Lieferant: ENZO LIFE SCIENCES
BMLP1990010
ENZOBMLP1990010EA
161
EUR
ENZOBMLP1990010
Z-VVR-AMC
Proteine und Peptide
Substrate for cathepsin S and cathepsin L
Sensitive fluorogenic substrate for cathepsin S and cathepsin L. Also cleaved by cathepsin B, cathepsin V, kallikrein 8, and factor VIIa/TF but not cathepsin K nor kallikrein 2. Ex.: 380 nm, Em.: 460 nm, although the following Ex/Em can also be used: 355,375/440,450. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144).