Compound 48/80
Lieferant: ENZO LIFE SCIENCES
BMLP2270001
ENZOBMLP2270001EA 211 EUR
ENZOBMLP2270001
Compound 48/80
Proteine und Peptide
Protease substrate
Fluorogenic substrate for ECE-1 and ECE-2, ACE1 and ACE2, neprilysin, MMP-2 and MMP-9, thimet oligopeptidase, as well as cathepsin A, cathepsin X/Z, BACE, and insulinase, but not MMP-1.Mca fluorescence is quenched by the Dnp group until cleavage separates them. Ex.: 328 nm, Em.: 393 nm, although the following Ex/Em have also been used: 320-340/400-420. This substrate is useful for inhibitor screening and kinetic analysis.