Suc-Ala-Ala-Pro-Phe-pNA
Lieferant: ENZO LIFE SCIENCES
BMLP2290010
ENZOBMLP2290010EA 238 EUR
ENZOBMLP2290010
Suc-Ala-Ala-Pro-Phe-pNA
Proteine und Peptide
Cathepsin substrate
Sensitive fluorogenic substrate for cathepsin K (Km=8µM, kcat/Km=4×10 M-1s-1). This substrate is also cleaved by cathepsin B, F, L, L2/V, S. Z-LR-AMC is also cleaved by malaria parasite cysteine proteases falcipain-1, -2, -3, berghepain, and vivapain-2, -3. Ex.: 365nm, Em.: 440nm, although the following Ex/Em can also be used: 355,380/450,460. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144).