SAMS Peptide
Lieferant: ENZO LIFE SCIENCES
BMLP2310001
ENZOBMLP2310001EA
162
EUR
ENZOBMLP2310001
SAMS Peptide
Proteine und Peptide
AMPK substrate
This peptide, HMRSAMSGLHLVKRR, is derived from rat acetyl-CoA carboxylase and is known to be a good substrate for AMPK.