H-Met-AMC ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: L-Methionine 7-amido-4-methylcoumarin trifluoroacetate salt

BMLP2360025
ENZOBMLP2360025EA 139 EUR
ENZOBMLP2360025
H-Met-AMC ≥98% (durch TLC)
H-Met-AMC

Fluorogenic substrate for methionine aminopeptidase 2 (MetAP2) (kcat/Km=1.1 x 10 M-1min-1; Km=310 µM) or aminopeptidase N (Km=377 µM). Ex.: 380 nm, Em.: 460 nm, although the following Ex/Em can also be used: 355,375/440,450. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144).


  • Aminopeptidase substrate


White to off white powder

Formel: C₁₅H₁₈N₂O₃S·C₂HF₃O₂
Molecular Weight: 420.40 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00210065
CAS-Nummer: 94367-35-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR