VEGFR-2 Substrate (Biotin)
Lieferant: ENZO LIFE SCIENCES
BMLP2410001
ENZOBMLP2410001EA
182
EUR
ENZOBMLP2410001
VEGFR-2 Substrate (Biotin)
Proteine und Peptide
This peptide, Biotin-Ahx-EEEEAYGWLDF, is derived from gastrin and can be used as a substrate for KDR/VEGFR2.
The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes.