FGFR substrate,Biotin
Lieferant: ENZO LIFE SCIENCES
BMLP2450001
ENZOBMLP2450001EA 234 EUR
ENZOBMLP2450001
FGFR substrate,Biotin
Proteine und Peptide
This peptide, Biotin-Ahx-KEAPEDLYKDFLTL, is derived from the KDR sequence and is a substrate for FGFR.
The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes.