Axltide, biotin
Lieferant: ENZO LIFE SCIENCES
BMLP2490001
ENZOBMLP2490001EA 182 EUR
ENZOBMLP2490001
Axltide, biotin
Proteine und Peptide
Protein kinase substrate
This peptide, biotin-Ahx-KKSRGDYMTMQIG, is derived from mouse insulin receptor 1 and is a substrate for Axl, DDR2, Mst1, and JAK2 kinases. The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes.