CDK2 substrate,Biotin
Lieferant: ENZO LIFE SCIENCES
BMLP2660001
ENZOBMLP2660001EA 136 EUR
ENZOBMLP2660001
CDK2 substrate,Biotin
Proteine und Peptide
This peptide, Biotin-Axh-HHASPRK, is a specific substrate for CDK2. The biotin allows the peptide to be used in kinase assays with streptavidin-bound membranes.