LRRKtide, Biotin
Lieferant: ENZO LIFE SCIENCES
BMLP2680001
ENZOBMLP2680001EA
136
EUR
ENZOBMLP2680001
LRRKtide, Biotin
Proteine und Peptide
LRRK substrate
This peptide, Biotin-Ahx-RLGRDKYKTLRQIRQ, is derived from ezrin/moesin/radixin and is a substrate for LRRK kinase. The biotin allows the peptide to be used in kinase assays with streptavidin-bound membranes.