CdC2 Substrate (Biotin)
Lieferant: ENZO LIFE SCIENCES
BMLP2720001
ENZOBMLP2720001EA 165 EUR
ENZOBMLP2720001
CdC2 Substrate (Biotin)
Proteine und Peptide
This peptide, (Ac-S)-PGRRRRK-biotin, is a substrate for CDC2 kinase.