Z-VAD(OMe)-FMK
Lieferant: ENZO LIFE SCIENCES
BMLP4160001
ENZOBMLP4160001EA 173 EUR
ENZOBMLP4160001
Z-VAD(OMe)-FMK
Proteine und Peptide
Caspase inhibitor
Irreversible and cell permeable broad-spectrum caspase inhibitor. Inhibits Fas-mediated apoptosis in Jurkat T cells and apoptosis in THP.1 cells induced by diverse stimuli (including cycloheximide, thapsigargin, etoposide and staurosporine). For cell free assays use Prod. No. ALX-260-020.