Ac-VEID-pNA
Lieferant: ENZO LIFE SCIENCES
BMLP4370005
ENZOBMLP4370005EA
141
EUR
ENZOBMLP4370005
Ac-VEID-pNA
Proteine und Peptide
Caspase substrate
Colorimetric substrate for caspase-6 (Km=30 µM; Kcat/Km=168’000 M-1sec-1) and related cysteine proteases. Sequence is based on the lamin A site of cleavage. Cleavage of the substrate by a caspase releases pNA, which can be detected by its absorbance at 405 nm (ε=10,500 M-1 cm-1).