Ac-REQD-pNA
Lieferant: ENZO LIFE SCIENCES
BMLP4420005
ENZOBMLP4420005EA
161
EUR
ENZOBMLP4420005
Ac-REQD-pNA
Proteine und Peptide
Caspase substrate
Colorimetric caspase substrate. Sequence corresponds to the prodomain of procaspase-8 cleavage site, residues 213-216, a putative caspase-10 target. Selective caspase-1 substrate at concentrations of 0,2 to 2 mM.