VP-AMC
Lieferant: ENZO LIFE SCIENCES
BMLP4480005
ENZOBMLP4480005EA
110
EUR
ENZOBMLP4480005
VP-AMC
Proteine und Peptide
DPP substrate
Fluorogenic substrate likely cleaved by dipeptidyl peptidase IV (DPIV, DPPIV, CD26), DPP8, FAP (fibroblast activation protein), and other prolyl peptidases. Can be used in vitro, for inhibitor screening, and in biological fluids. Ex = 380 nm, Em = 460 nm.