WP-AMC
Lieferant: ENZO LIFE SCIENCES
BMLP4500005
ENZOBMLP4500005EA
136
EUR
ENZOBMLP4500005
WP-AMC
Proteine und Peptide
DPP substrate
Fluorogenic substrate likely cleaved by dipeptidyl peptidase IV (DPIV, DPPIV, CD26), DPP8, and other prolyl peptidases. Can be used in vitro, for inhibitor screening, and in biological fluids. Ex = 380 nm, Em = 460 nm.