IKK-NBD control peptide
Lieferant: ENZO LIFE SCIENCES
BMLP6080500
ENZOBMLP6080500EA 212 EUR
ENZOBMLP6080500
IKK-NBD control peptide
Proteine und Peptide
Negative control for IKK-NBD peptide
Negative control peptide for IKK-NBD peptide. Sold under license from Vanderbilt University