Suc-Leu-Leu-Val-Tyr-AMC
Lieferant: ENZO LIFE SCIENCES
BMLP8020005
ENZOBMLP8020005EA 114 EUR
ENZOBMLP8020005
Suc-Leu-Leu-Val-Tyr-AMC
Proteine und Peptide
Proteasome substrate
Fluorogenic substrate for 20S proteasome (Prod. No. BML-PW8720) and other chymotrypsin-like proteases, as well as calpains. Ex.: 380 nm Em.: 460 nm.