EHNA ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Erythro-9-(2-hydroxy-3-nonyl)adenine hydrochloride

BMLPD1290010 BMLPD1290050
ENZOBMLPD1290010EA 68.2 EUR
ENZOBMLPD1290010 ENZOBMLPD1290050
EHNA ≥98% (durch HPLC)
EHNA

Phosphodiesterase PDE II inhibitor, IC50=0.8 µM. Also inhibits adenosine deaminase, Ki=4 nM.


  • PDE inhibitor


White solid.


Soluble in water (25mg/ml).

Formel: C₁₄H₂₃N₅O·HCl
Molecular Weight: 313.8 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 51350-19-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58a71c6af5c7fe67d0d2afcc08f30b0c4e1f701e&vid=e8fc146503bcaa0828b3cf7afed1fbcc327b6db1&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR