Irsogladin-Maleat ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 6-(2,5-Dichlorophenyl)-1,3,5-triazine-2,4-diamine maleate, 2,4-Diamino-6-(2,5-dichlorophenyl)-1,3,5-triazine Maleate

BMLPD1410050
ENZOBMLPD1410050EA 84.7 EUR
ENZOBMLPD1410050
Irsogladin-Maleat ≥98%
Irsogladine maleate

PDE inhibitor


Irsogladine is a selective PDE4 inhibitor which also enhances gap junction intercellular communication. It inhibits angiogenesis in wild-type and plasminogen activator-deficient mice.

Formel: C₉H₇Cl₂N₅
Molecular Weight: 372,16 g/mol
MDL: MFCD00873566
CAS-Nummer: 84504-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58d50d307ec891e2ab17f98cb7d543707a71ec7a&vid=18287ac0d66f32f04546bfb97e24547d34d8303b&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR