8-Methoxymethyl-IBMX ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
BMLPD1500010
ENZOBMLPD1500010EA
234
EUR
ENZOBMLPD1500010
8-Methoxymethyl-IBMX ≥98% (durch TLC)
8-Methoxymethyl-IBMX
Selective inhibitor of Ca2+-calmodulin-dependent phosphodiesterase (PDE I, IC50=4 µM).
- PDE inhibitor
White solid.
Soluble in 100% ethanol (5mg/ml) or DMSO (5mg/ml)
Formel:
C₁₂H₁₈N₄O₃ Molecular Weight: 266.3 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
78033-08-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...