R-(-)-Rolipram ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: (R)-(-)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]pyrrolidin-2-one, R-(-)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]-2-pyrrolidinone

BMLPD1770005
ENZOBMLPD1770005EA 236 EUR
ENZOBMLPD1770005
R-(-)-Rolipram ≥98% (durch HPLC)
R-(-)-Rolipram

R-enantiomer of rolipram which is more active at inhibition of PDE IV, Ki=06 µM. 10-fold more active than S-rolipram at inhibition of PDE IV. Inhibits antigen-induced contraction of guinea-pig colonic smooth muscle, EC50=0.1 µM.


  • PDE inhibitor


White to off-white solid.


Soluble in DMSO.

Formel: C₁₆H₂₁NO₃
Molecular Weight: 275,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03093860
CAS-Nummer: 85416-75-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5646573eb53e74566d8f71abbc6cb7a8cf864389&vid=8565f4ca38838e5f7bfeaf4675caa9219acc773d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR